Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GXYLT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193691.3 | NP_001180620.1 | 474 | 81 | ||
XM_028843068.1 | XP_028698901.1 | 632 | 81 | |||
GenBank mRNA | JV070198.1 | - | 280 | 81 |