Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261012.2 | NP_001247941.1 | 813 | 60 | ||
XM_015149721.2 | XP_015005207.2 | 1029 | 60 | |||
XM_015149722.2 | XP_015005208.2 | 1050 | 60 | |||
XM_015149723.2 | XP_015005209.2 | 867 | 60 | |||
XM_015149724.2 | XP_015005210.2 | 921 | 60 | |||
XM_015149725.2 | XP_015005211.2 | 877 | 60 | |||
XM_028827345.1 | XP_028683178.1 | 1122 | 60 | |||
XM_028827346.1 | XP_028683179.1 | 908 | 60 | |||
XM_028827347.1 | XP_028683180.1 | 943 | 60 | |||
XM_028827348.1 | XP_028683181.1 | 870 | 60 | |||
GenBank mRNA | JU474331.1 | - | 832 | 60 | ||
JV046207.1 | - | 817 | 60 | |||
JV457071.1 | - | 829 | 60 | |||
JV489111.1 | - | 829 | 60 | |||
JV673111.1 | - | 821 | 60 |
Gene Symbol: ARVCF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JU325911.1 | - | 4917 | 60 | ||
JV582160.1 | - | 4027 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Macaca mulatta |