Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261480.1 | NP_001248409.1 | 990 | 114 | ||
GenBank mRNA | JU326985.1 | - | 336 | 114 | ||
JU477353.1 | - | 990 | 114 | |||
JV594909.1 | - | 639 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |