Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eda | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006257110.2 | XP_006257172.1 | 6 - 7 | 993 | 68 | |
XM_006257111.2 | XP_006257173.1 | 6 - 7 | 993 | 68 | ||
XM_008773301.1 | XP_008771523.1 | 6 - 7 | 993 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Rattus norvegicus Probe spans exons |