Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnih2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025132.1 | NP_001020303.1 | 4 - 5 | 559 | 92 | |
XM_008760153.1 | XP_008758375.1 | 3 - 4 | 1137 | 92 | ||
XM_008760154.1 | XP_008758376.1 | 4 - 5 | 1197 | 92 | ||
GenBank mRNA | BC091325.1 | - | 4 - 5 | 559 | 7313787 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |