Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013094.1 | NP_037226.1 | 3 - 4 | 420 | 81 | |
XM_008759499.1 | XP_008757721.1 | 4 - 5 | 588 | 81 | ||
GenBank mRNA | L26043.1 | - | 3 - 4 | 420 | 81 | |
L26044.1 | - | 3 - 4 | 420 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |