Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oxt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012996.3 | NP_037128.1 | 2 - 3 | 356 | 78 | |
GenBank mRNA | M25649.1 | - | 2 - 3 | 356 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Probe spans exons |