Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gja6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019308.1 | NP_062181.1 | 1 | 360 | 67 | |
XM_006256894.2 | XP_006256956.1 | 2 | 737 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Inventoried Both primers and probe map within a single exon |