Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Calr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022399.2 | NP_071794.1 | 7 - 8 | 1028 | 91 | |
GenBank mRNA | BC062395.1 | - | 7 - 8 | 1005 | 5623178 | 91 |
D78308.1 | - | 7 - 8 | 980 | 91 | ||
FQ210587.1 | - | 1033 | 91 | |||
FQ226996.1 | - | 1 - 2 | 113 | 91 | ||
X13702.1 | - | 1 - 2 | 161 | 91 | ||
X53363.1 | - | 7 - 8 | 1000 | 91 | ||
X79327.1 | - | 7 - 8 | 978 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Rat Androgen Panel 1700 Inventoried Probe spans exons |