Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Doc2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022937.2 | NP_075226.1 | 3 - 4 | 487 | 82 | |
XM_006230335.2 | XP_006230397.1 | 4 - 5 | 587 | 82 | ||
XM_006230336.1 | XP_006230398.1 | 4 - 5 | 771 | 82 | ||
XM_006230337.2 | XP_006230399.1 | 4 - 5 | 583 | 82 | ||
XM_006230339.2 | XP_006230401.1 | 4 - 5 | 521 | 82 | ||
XM_006230340.1 | XP_006230402.1 | 4 - 5 | 705 | 82 | ||
XM_006230341.2 | XP_006230403.1 | 4 - 5 | 705 | 82 | ||
XM_008759918.1 | XP_008758140.1 | 4 - 5 | 618 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Probe spans exons |