Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slco1a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030838.1 | NP_110465.1 | 1 - 2 | 184 | 118 | |
GenBank mRNA | AF083469.1 | - | 1 - 2 | 184 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Rattus norvegicus Inventoried Probe spans exons |