Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lypla2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031342.1 | NP_112632.1 | 4 - 5 | 264 | 78 | |
GenBank mRNA | AB021645.1 | - | 4 - 5 | 264 | 78 | |
BC070503.1 | - | 4 - 5 | 270 | 7096587 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |