Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ghsr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | U94321.1 | - | 1 | 849 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Inventoried Both primers and probe map within a single exon |