Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131908.1 | NP_571983.1 | 2 - 3 | 456 | 106 | |
GenBank mRNA | AB079674.1 | - | 2 - 3 | 456 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |