Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr1d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_147210.2 | NP_671743.2 | 5 - 6 | 1346 | 100 | |
GenBank mRNA | U15660.1 | - | 4 - 5 | 1063 | 100 | |
U20796.1 | - | 4 - 5 | 1003 | 100 | ||
X82777.1 | - | 5 - 6 | 1281 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Nuclear Receptor Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |