Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parva | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020656.2 | NP_065707.2 | 5 - 6 | 548 | 59 | |
GenBank mRNA | AF264765.2 | - | 5 - 6 | 548 | 59 | |
FQ228521.1 | - | 608 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |