Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024373.1 | NP_077349.1 | 1 - 2 | 244 | 101 | |
XM_006254471.2 | XP_006254533.1 | 2 - 3 | 1665 | 101 | ||
GenBank mRNA | U35890.1 | - | 1 - 2 | 244 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |