Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031119.2 | NP_112381.2 | 8 - 9 | 771 | 88 | |
XM_006234321.2 | XP_006234383.1 | 8 - 9 | 847 | 88 | ||
XM_006234322.2 | XP_006234384.1 | 8 - 9 | 847 | 88 | ||
XM_006234323.2 | XP_006234385.1 | 8 - 9 | 847 | 88 | ||
GenBank mRNA | BC081780.1 | - | 8 - 9 | 771 | 7192454 | 88 |
BC097252.1 | - | 7 - 8 | 851 | 88 | ||
X67859.1 | - | 7 - 8 | 706 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |