Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053511.1 | NP_445963.1 | 2 - 3 | 468 | 102 | |
XM_008764298.1 | XP_008762520.1 | 3 - 4 | 671 | 102 | ||
GenBank mRNA | AF098301.1 | - | 2 - 3 | 468 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |