Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008759812.1 | XP_008758034.1 | 10 - 11 | 2185 | 98 | |
GenBank mRNA | AF136401.1 | - | 2192 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Ion Channel Rattus norvegicus Probe spans exons |