Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncdn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053543.2 | NP_445995.2 | 6 - 7 | 2113 | 51 | |
XM_006238908.1 | XP_006238970.1 | 6 - 7 | 1822 | 51 | ||
XM_006238909.1 | XP_006238971.1 | 6 - 7 | 1883 | 51 | ||
GenBank mRNA | BC087000.1 | - | 6 - 7 | 2113 | 7190905 | 51 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |