Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkar1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013181.1 | NP_037313.1 | 10 - 11 | 1074 | 95 | |
XM_008768311.1 | XP_008766533.1 | 10 - 11 | 1130 | 95 | ||
GenBank mRNA | M17086.1 | - | 10 - 11 | 1074 | 95 | |
U75932.1 | - | 7 - 8 | 588 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Rattus norvegicus 1700 Probe spans exons |