Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153629.1 | NP_705893.1 | 7 - 8 | 1131 | 112 | |
XM_006246232.1 | XP_006246294.1 | 7 - 8 | 1175 | 112 | ||
GenBank mRNA | AF077354.1 | - | 7 - 8 | 1131 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |