Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fads3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173137.1 | NP_775160.1 | 7 - 8 | 989 | 111 | |
GenBank mRNA | AJ494720.1 | - | 7 - 8 | 989 | 111 | |
HQ901598.1 | - | 7 - 8 | 913 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |