Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114330.1 | NP_001107802.1 | 2 - 3 | 1078 | 77 | |
NM_017011.1 | NP_058707.1 | 2 - 3 | 1078 | 77 | ||
GenBank mRNA | M61099.1 | - | 2 - 3 | 1078 | 77 | |
X57569.1 | - | 2 - 3 | 1060 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Rattus norvegicus Probe spans exons |