Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfpi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017200.1 | NP_058896.1 | 7 - 8 | 913 | 130 | |
XM_006234440.2 | XP_006234502.1 | 8 - 9 | 1208 | 130 | ||
XM_006234441.2 | XP_006234503.1 | 8 - 9 | 2229 | 130 | ||
XM_006234444.2 | XP_006234506.1 | 8 - 9 | 2242 | 130 | ||
XM_008761980.1 | XP_008760202.1 | 8 - 9 | 1227 | 130 | ||
XM_008761981.1 | XP_008760203.1 | 8 - 9 | 991 | 130 | ||
XM_008761982.1 | XP_008760204.1 | 8 - 9 | 1221 | 130 | ||
XM_008761983.1 | XP_008760205.1 | 8 - 9 | 2244 | 130 | ||
XM_008761984.1 | XP_008760206.1 | 8 - 9 | 2257 | 130 | ||
GenBank mRNA | D10926.1 | - | 7 - 8 | 913 | 130 | |
FQ219876.1 | - | 5 - 6 | 619 | 130 | ||
FQ220339.1 | - | 5 - 6 | 613 | 130 | ||
FQ220655.1 | - | 5 - 6 | 710 | 130 | ||
FQ229364.1 | - | 5 - 6 | 642 | 130 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Rattus norvegicus 1700 Probe spans exons |