Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Espn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006239476.2 | XP_006239538.1 | 10 - 11 | 2339 | 104 | |
XM_008764375.1 | XP_008762597.1 | 10 - 11 | 2348 | 104 | ||
XM_008764377.1 | XP_008762599.1 | 9 - 10 | 2300 | 104 | ||
XM_008764379.1 | XP_008762601.1 | 11 - 12 | 2258 | 104 | ||
XM_008764380.1 | XP_008762602.1 | 10 - 11 | 2210 | 104 | ||
GenBank mRNA | AF540946.1 | - | 5 - 6 | 1030 | 104 | |
AF540947.1 | - | 6 - 7 | 1057 | 104 | ||
AF540948.1 | - | 4 - 5 | 802 | 104 | ||
AF540949.1 | - | 5 - 6 | 829 | 104 | ||
AY587568.1 | - | 3 - 4 | 389 | 104 | ||
AY587569.1 | - | 2 - 3 | 479 | 104 | ||
U46007.1 | - | 10 - 11 | 2024 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons |