Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Erc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_170787.2 | NP_740768.1 | 9 - 10 | 1920 | 104 | |
GenBank mRNA | AF541925.1 | - | 9 - 10 | 1920 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |