Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cct4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182814.2 | NP_877966.1 | 12 - 13 | 1623 | 88 | |
GenBank mRNA | AY223861.1 | - | 12 - 13 | 1489 | 88 | |
BC079283.1 | - | 12 - 13 | 1594 | 7134210 | 88 | |
BQ200856.1 | - | 2 - 3 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |