Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nampt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177928.3 | NP_808789.1 | 6 - 7 | 768 | 112 | |
GenBank mRNA | AB081730.1 | - | 6 - 7 | 743 | 112 | |
AY831728.1 | - | 6 - 7 | 741 | 112 | ||
BC085681.1 | - | 6 - 7 | 768 | 7192109 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |