Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sftpa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270645.1 | NP_001257574.1 | 3 - 4 | 353 | 58 | |
NM_001270647.1 | NP_001257576.1 | 3 - 4 | 564 | 58 | ||
NM_017329.2 | NP_059025.2 | 3 - 4 | 350 | 58 | ||
GenBank mRNA | BC085353.1 | - | 3 - 4 | 353 | 7121530 | 58 |
CK364490.1 | - | 3 - 4 | 576 | 58 | ||
M15754.1 | - | 3 - 4 | 338 | 58 | ||
M33201.1 | - | 3 - 4 | 347 | 58 | ||
X13176.1 | - | 3 - 4 | 338 | 58 | ||
X13177.1 | - | 3 - 4 | 347 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |