Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsks | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199088.1 | NP_954519.1 | 9 - 10 | 1504 | 70 | |
XM_006229017.2 | XP_006229079.1 | 9 - 10 | 1538 | 70 | ||
XM_006229019.2 | XP_006229081.1 | 9 - 10 | 1565 | 70 | ||
XM_006229020.2 | XP_006229082.1 | 9 - 10 | 1538 | 70 | ||
GenBank mRNA | AY346104.1 | - | 9 - 10 | 1504 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Rattus norvegicus Probe spans exons |