Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2rx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270621.1 | NP_001257550.1 | 5 - 6 | 673 | 64 | |
NM_031075.2 | NP_112337.2 | 6 - 7 | 745 | 64 | ||
NR_073054.1 | 7 - 8 | 767 | 64 | |||
GenBank mRNA | AF084975.1 | - | 5 - 6 | 518 | 64 | |
BC081783.1 | - | 7 - 8 | 767 | 7134300 | 64 | |
X90651.1 | - | 6 - 7 | 745 | 64 | ||
X91167.1 | - | 6 - 7 | 735 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Mammalian Gene Collection Rattus norvegicus Probe spans exons |