Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnpo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100692.1 | NP_001094162.1 | 19 - 20 | 2328 | 62 | |
XM_006231839.2 | XP_006231901.1 | 20 - 21 | 2919 | 62 | ||
XM_006231840.2 | XP_006231902.1 | 20 - 21 | 2643 | 62 | ||
XM_008760668.1 | XP_008758890.1 | 20 - 21 | 2931 | 62 | ||
GenBank mRNA | BC090323.1 | - | 17 - 18 | 2008 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |