Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tatdn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105987.1 | NP_001099457.1 | 2 - 3 | 140 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |