Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zpbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025139.1 | NP_001020310.1 | 2 - 3 | 459 | 68 | |
XM_006251505.2 | XP_006251567.1 | 2 - 3 | 489 | 68 | ||
GenBank mRNA | BC079315.1 | - | 2 - 3 | 459 | 7134936 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |