Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ikzf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106916.1 | NP_001100386.1 | 6 - 7 | 865 | 66 | |
XM_006245119.2 | XP_006245181.1 | 6 - 7 | 1011 | 66 | ||
XM_006245120.2 | XP_006245182.1 | 6 - 7 | 759 | 66 | ||
XM_006245121.2 | XP_006245183.1 | 6 - 7 | 906 | 66 | ||
XM_008767217.1 | XP_008765439.1 | 6 - 7 | 860 | 66 | ||
XM_008767218.1 | XP_008765440.1 | 6 - 7 | 838 | 66 | ||
XM_008767219.1 | XP_008765441.1 | 6 - 7 | 797 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Rattus norvegicus Probe spans exons |