Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lztfl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024266.1 | NP_001019437.1 | 4 - 5 | 433 | 98 | |
XM_006244191.1 | XP_006244253.1 | 4 - 5 | 577 | 98 | ||
GenBank mRNA | BC092585.1 | - | 4 - 5 | 433 | 7127181 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |