Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4galt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031740.1 | NP_113928.1 | 4 - 5 | 1313 | 69 | |
GenBank mRNA | AF048687.1 | - | 4 - 5 | 1313 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |