Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syt10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031666.1 | NP_113854.1 | 2 - 3 | 509 | 60 | |
GenBank mRNA | AF375463.1 | - | 2 - 3 | 509 | 60 | |
DQ437524.1 | - | 2 - 3 | 509 | 60 | ||
U85513.1 | - | 2 - 3 | 641 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |