Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108103.1 | NP_001101573.1 | 8 - 9 | 2519 | 71 | |
XM_006242194.2 | XP_006242256.1 | 8 - 9 | 2908 | 71 | ||
XM_006242196.2 | XP_006242258.1 | 8 - 9 | 2515 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |