Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gxylt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100887.1 | NP_001094357.1 | 6 - 7 | 1027 | 134 | |
XM_006242192.2 | XP_006242254.1 | 5 - 6 | 935 | 134 | ||
GenBank mRNA | AY623036.1 | - | 4 - 5 | 630 | 134 | |
BC167068.1 | - | 5 - 6 | 781 | 8363586 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |