Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmg20b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108731.1 | NP_001102201.1 | 5 - 6 | 823 | 120 | |
XM_006240944.2 | XP_006241006.1 | 6 - 7 | 720 | 120 | ||
XM_006240946.2 | XP_006241008.1 | 6 - 7 | 642 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |