Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006996.1 | NP_001006997.1 | 7 - 8 | 1245 | 89 | |
GenBank mRNA | BC083919.1 | - | 7 - 8 | 1245 | 7135005 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |