Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nckap5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008769508.1 | XP_008767730.1 | 2 - 3 | 459 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |