Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc18a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013031.1 | NP_037163.1 | 4 - 5 | 628 | 82 | |
XM_006231664.2 | XP_006231726.1 | 4 - 5 | 1469 | 82 | ||
XM_008760515.1 | XP_008758737.1 | 3 - 4 | 828 | 82 | ||
XM_008760516.1 | XP_008758738.1 | 4 - 5 | 1474 | 82 | ||
XM_008760517.1 | XP_008758739.1 | 4 - 5 | 1469 | 82 | ||
XM_008760518.1 | XP_008758740.1 | 3 - 4 | 828 | 82 | ||
XM_008760519.1 | XP_008758741.1 | 3 - 4 | 828 | 82 | ||
GenBank mRNA | L00603.1 | - | 4 - 5 | 628 | 82 | |
M97381.1 | - | 4 - 5 | 599 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Detect Genomic DNA Rattus norvegicus Probe spans exons |