Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdhc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005534.1 | NP_001005534.1 | 5 - 6 | 417 | 81 | |
XM_008769731.1 | XP_008767953.1 | 6 - 7 | 464 | 81 | ||
GenBank mRNA | BC082027.1 | - | 5 - 6 | 417 | 7130052 | 81 |
FQ210665.1 | - | 5 - 6 | 419 | 81 | ||
FQ213406.1 | - | 5 - 6 | 436 | 81 | ||
FQ215668.1 | - | 6 - 7 | 471 | 81 | ||
FQ215689.1 | - | 5 - 6 | 419 | 81 | ||
FQ215712.1 | - | 5 - 6 | 419 | 81 | ||
FQ216122.1 | - | 5 - 6 | 435 | 81 | ||
FQ216612.1 | - | 5 - 6 | 436 | 81 | ||
FQ216658.1 | - | 5 - 6 | 419 | 81 | ||
FQ216892.1 | - | 5 - 6 | 435 | 81 | ||
FQ217679.1 | - | 5 - 6 | 436 | 81 | ||
FQ218940.1 | - | 5 - 6 | 436 | 81 | ||
FQ225195.1 | - | 5 - 6 | 430 | 81 | ||
FQ229651.1 | - | 5 - 6 | 436 | 81 | ||
FQ231805.1 | - | 5 - 6 | 436 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |