Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lin9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001067807.5 | XP_001067807.3 | 10 - 11 | 1321 | 120 | |
XM_006250417.1 | XP_006250479.1 | 11 - 12 | 1160 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |