Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Birc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008762598.1 | XP_008760820.1 | 5 - 6 | 1528 | 91 | |
XM_008762599.1 | XP_008760821.1 | 5 - 6 | 1528 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Apoptosis 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons |