Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013131.1 | NP_001013149.1 | 3 - 4 | 416 | 69 | |
GenBank mRNA | BC091207.1 | - | 3 - 4 | 416 | 7381272 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |